Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-iodobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

23831-1G 23831-25G 23831-500MG 23831-5G
76911-396EA 198.39 USD InStock
76911-396 76911-398 76911-400 76911-402
3,5-Dichloro-4-iodobenzoic acid ≥97% research grade
3,5-Dichloro-4-iodobenzoic acid


Formula: C₇H₃Cl₂IO₂
MW: 316.908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14704259
CAS Number: 117757-68-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty