Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloro-2-isopropoxypyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

24454-1G 24454-25G 24454-500MG 24454-5G
76911-618EA 307.67 USD InStock
76911-618 76911-620 76911-622 76911-624
3-Bromo-5-chloro-2-isopropoxypyridine ≥97% research grade
3-Bromo-5-chloro-2-isopropoxypyridine


Formula: C₈H₉BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD22628195
CAS Number: 1289133-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty