Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminocarbonyl)-2-chlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

24540-1G 24540-25G 24540-500MG 24540-5G
76911-656EA 337.37 USD InStock
76911-656 76911-658 76911-660 76911-662
4-(Aminocarbonyl)-2-chlorophenylboronic acid ≥97%
4-(Aminocarbonyl)-2-chlorophenylboronic acid


Formula: C₇H₇BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD31619415
CAS Number: 2691129-23-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty