Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylthio)-5-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

25731-1G 25731-25G 25731-500MG 25731-5G
76912-044EA 341.61 USD InStock
76912-044 76912-046 76912-048 76912-050
3-(Methylthio)-5-(trifluoromethyl)benzoic acid ≥97% research grade
3-(Methylthio)-5-(trifluoromethyl)benzoic acid


Formula: C₉H₇F₃O₂S
MW: 236.211 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22488997
CAS Number: 53985-40-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty