Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ffluoro-N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥97%

Supplier: AOB CHEM USA
Warning

26918-1G 26918-25G 26918-500MG 26918-5G
76912-572EA 290.69 USD InStock
76912-572 76912-574 76912-576 76912-578
3-Ffluoro-N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥97%
3-Ffluoro-N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline


Formula: C₁₄H₂₁BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD22495043
CAS Number: 1129542-03-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty