Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-6-(methylthio)pyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

27484-1G 27484-25G 27484-500MG 27484-5G
76912-820EA 225.98 USD InStock
76912-820 76912-822 76912-824 76912-826
3-Bromo-2-chloro-6-(methylthio)pyridine ≥97% research grade
3-Bromo-2-chloro-6-(methylthio)pyridine


Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28098203
CAS Number: 1809158-14-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty