Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclobutylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

28880-1G 28880-25G 28880-500MG 28880-5G
76913-080EA 290.69 USD InStock
76913-080 76913-082 76913-084 76913-086
3-Cyclobutylphenylboronic acid ≥97%
3-Cyclobutylphenylboronic acid


Formula: C₁₀H₁₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12760761
CAS Number: 1381885-38-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty