Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[3-(Benzyloxy)-4-methoxyphenyl]methanol ≥95% research grade

Supplier: AOB CHEM USA
Warning

29189-1G 29189-25G 29189-500MG 29189-5G
76913-182EA 162.32 USD InStock
76913-182 76913-184 76913-186 76913-188
[3-(Benzyloxy)-4-methoxyphenyl]methanol ≥95% research grade
[3-(Benzyloxy)-4-methoxyphenyl]methanol


Formula: C₁₅H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00183283
CAS Number: 1860-60-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty