Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoro-2-methylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

29279-1G 29279-25G 29279-5G
76913-238EA 114.58 USD InStock
76913-238 76913-240 76913-242
3,4-Difluoro-2-methylbenzaldehyde ≥97% research grade
3,4-Difluoro-2-methylbenzaldehyde


Formula: C₈H₆F₂O
MW: 156.129 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07777151
CAS Number: 847502-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty