Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

29418-1G 29418-500MG 29418-5G
76913-278EA 683.22 USD InStock
76913-278 76913-280 76913-282
3-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester ≥97% research grade
3-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester


Formula: C₁₈H₂₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD24449435
CAS Number: 1366131-49-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty