Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isopropylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

29969-25G 29969-5G
76913-458EA 545.31 USD InStock
76913-458 76913-460
3-Isopropylphenylboronic acid pinacol ester ≥97%
3-Isopropylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD13195750
CAS Number: 325142-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty