Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloroisonicotinaldehyde hydrochloride ≥97%

Supplier: AOB CHEM USA
Warning

30082-25G 30082-5G
76913-470EA 275.84 USD InStock
76913-470 76913-472
3-Chloroisonicotinaldehyde hydrochloride ≥97%
3-Chloroisonicotinaldehyde hydrochloride


Formula: C₆H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD28134601
CAS Number: 1826110-00-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty