Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Tert-butoxycarbonyl)-3-methylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

30154-1G 30154-500MG 30154-5G
76913-480EA 524.09 USD InStock
76913-480 76913-482 76913-484
4-(Tert-butoxycarbonyl)-3-methylphenylboronic acid pinacol ester ≥97% research grade
4-(Tert-butoxycarbonyl)-3-methylphenylboronic acid pinacol ester


Formula: C₁₈H₂₇BO₄
Storage Temperature: Ambient
CAS Number: 1331887-05-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty