Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-phenoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

30585-1G 30585-25G 30585-500MG 30585-5G
76913-580EA 352.22 USD InStock
76913-580 76913-582 76913-584 76913-586
3-Chloro-4-phenoxyphenylboronic acid ≥97%
3-Chloro-4-phenoxyphenylboronic acid


Formula: C₁₂H₁₀BClO₃
Storage Temperature: Ambient
MDL Number: MFCD29088166
CAS Number: 2095461-96-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty