Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,6-dimethoxy-5-methylpyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

30751-1G 30751-25G 30751-5G
76913-622EA 103.97 USD InStock
76913-622 76913-624 76913-626
3-Bromo-2,6-dimethoxy-5-methylpyridine ≥97% research grade
3-Bromo-2,6-dimethoxy-5-methylpyridine


Formula: C₈H₁₀BrNO₂
MW: 232.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088193
CAS Number: 145916-05-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty