Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-4-methylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

30925-1G 30925-25G 30925-500MG 30925-5G
76913-684EA 318.27 USD InStock
76913-684 76913-686 76913-688 76913-690
3,5-Difluoro-4-methylphenylboronic acid pinacol ester ≥97% research grade
3,5-Difluoro-4-methylphenylboronic acid pinacol ester


Formula: C₁₃H₁₇BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD29088222
CAS Number: 2094504-03-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty