Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

31108-1G 31108-25G 31108-500MG 31108-5G
76913-780EA 178.24 USD InStock
76913-780 76913-782 76913-784 76913-786
3,6-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97% research grade
3,6-Difluoro-2-methoxyphenylboronic acid pinacol ester


Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415003
CAS Number: 2121513-59-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty