Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-5-chloro-2-fluorophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

31206-1G 31206-500MG 31206-5G
76913-848EA 524.09 USD InStock
76913-848 76913-850 76913-852
4-Benzyloxy-5-chloro-2-fluorophenylboronic acid ≥97% research grade
4-Benzyloxy-5-chloro-2-fluorophenylboronic acid


Formula: C₁₃H₁₁BClFO₃
MW: 280.487 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088280
CAS Number: 2121512-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty