Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-3-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31468-1G 31468-25G 31468-5G
76913-980EA 88.06 USD InStock
76913-980 76913-982 76913-984
4-Acetyl-3-fluorobenzeneboronic acid pinacol ester ≥97%
4-Acetyl-3-fluorobenzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD18732911
CAS Number: 1351499-39-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty