Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formylthiophene-4-boronic acid pincol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

32314-1G 32314-25G 32314-500MG 32314-5G
76914-184EA 169.75 USD InStock
76914-184 76914-186 76914-188 76914-190
3-Formylthiophene-4-boronic acid pincol ester ≥97% research grade
3-Formylthiophene-4-boronic acid pincol ester


Formula: C₁₁H₁₅BO₃S
Storage Temperature: Ambient
MDL Number: MFCD18910605
CAS Number: 2121511-75-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty