Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

32406-1G 32406-25G 32406-500MG 32406-5G
76914-234EA 265.23 USD InStock
76914-234 76914-236 76914-238 76914-240
3-Chloro-5-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97% research grade
3-Chloro-5-(trifluoromethoxy)phenylboronic acid, pinacol ester


Formula: C₁₃H₁₅BClF₃O₃
MW: 322.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18756755
CAS Number: 1803320-97-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty