Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-(tetrahydro-pyran-2-yl)-5-trifluoromethyl-1H-indazole ≥95%

Supplier: AOB CHEM USA
Warning

32667-1G 32667-25G 32667-500MG 32667-5G
76914-432EA 361.77 USD InStock
76914-432 76914-434 76914-436 76914-438
4-Bromo-1-(tetrahydro-pyran-2-yl)-5-trifluoromethyl-1H-indazole ≥95%
4-Bromo-1-(tetrahydro-pyran-2-yl)-5-trifluoromethyl-1H-indazole


Formula: C₁₃H₁₂BrF₃N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD30609548
CAS Number: 2044704-81-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty