Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(pyrrolidin-1-yl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

32933-1G 32933-25G 32933-500MG 32933-5G
76914-494EA 376.62 USD InStock
76914-494 76914-496 76914-498 76914-500
3-Fluoro-5-(pyrrolidin-1-yl)phenol ≥97%
3-Fluoro-5-(pyrrolidin-1-yl)phenol


Formula: C₁₀H₁₂FNO
Storage Temperature: Refrigerator
MDL Number: MFCD23110831
CAS Number: 925233-15-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty