Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5-dichlorophenylboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

33671-1G 33671-25G 33671-500MG 33671-5G
76914-828EA 337.37 USD InStock
76914-828 76914-830 76914-832 76914-834
3-Bromo-2,5-dichlorophenylboronic acid ≥95% research grade
3-Bromo-2,5-dichlorophenylboronic acid


Formula: C₆H₄BBrCl₂O₂
MW: 269.716 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31382567
CAS Number: 2121513-17-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty