Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlor-2-methyl-phenol ≥95% research grade

Supplier: AOB CHEM USA
Warning

34013-1G 34013-500MG 34013-5G
76914-916EA 490.14 USD InStock
76914-916 76914-918 76914-920
3,5-Dichlor-2-methyl-phenol ≥95% research grade
3,5-Dichlor-2-methyl-phenol


Formula: C₇H₆Cl₂O
MW: 177.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415130
CAS Number: 56680-66-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty