Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-bromo-4-chloro-5-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

34110-1G 34110-25G 34110-500MG 34110-5G
76914-940EA 213.25 USD InStock
76914-940 76914-942 76914-944 76914-946
3-bromo-4-chloro-5-fluorobenzoic acid ≥97%
3-bromo-4-chloro-5-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21364899
CAS Number: 1357942-87-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty