Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoro-5-methylbenzonitrile ≥97% research grade

Supplier: AOB CHEM USA
Warning

34299-1G 34299-500MG 34299-5G
76915-024EA 537.88 USD InStock
76915-024 76915-026 76915-028
3-Bromo-2-fluoro-5-methylbenzonitrile ≥97% research grade
3-Bromo-2-fluoro-5-methylbenzonitrile


Formula: C₈H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD12756720
CAS Number: 1260741-40-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty