Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trichlorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34341-1G 34341-25G 34341-500MG 34341-5G
76915-060EA 132.62 USD InStock
76915-060 76915-062 76915-064 76915-066
3,4,5-Trichlorobenzoic acid ≥97% research grade
3,4,5-Trichlorobenzoic acid


Formula: C7H3Cl3O2
Storage Temperature: Ambient
MDL Number: MFCD00061128
CAS Number: 51-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty