Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-6-iodopyridin-3-ol ≥95% research grade

Supplier: AOB CHEM USA
Warning

34524-1G 34524-25G 34524-500MG 34524-5G
76915-118EA 222.79 USD InStock
76915-118 76915-120 76915-122 76915-124
4-Bromo-2-chloro-6-iodopyridin-3-ol ≥95% research grade
4-Bromo-2-chloro-6-iodopyridin-3-ol


Formula: C₅H₂BrClINO
MW: 334.337 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31805771
CAS Number: 2244107-68-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty