Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

34653-1G 34653-25G 34653-500MG 34653-5G
76915-144EA 239.77 USD InStock
76915-144 76915-146 76915-148 76915-150
3,4-Difluoro-2-methoxyphenylboronic acid pinacol ester ≥97% research grade
3,4-Difluoro-2-methoxyphenylboronic acid pinacol ester


Formula: C₁₃H₁₇BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22414983
CAS Number: 2230088-81-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty