Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

34769-1G 34769-25G 34769-500MG 34769-5G
76915-198EA 270.53 USD InStock
76915-198 76915-200 76915-202 76915-204
3,5-Dimethylphenylboronic acid pinacol ester ≥97% research grade
3,5-Dimethylphenylboronic acid pinacol ester


Formula: C₁₄H₂₀BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12405367
CAS Number: 1147894-98-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty