Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Bromophenyl)thiophene ≥97% research grade

Supplier: AOB CHEM USA
Warning

34856-1G 34856-25G 34856-500MG 34856-5G
76915-244EA 377.69 USD InStock
76915-244 76915-246 76915-248 76915-250
3-(4-Bromophenyl)thiophene ≥97% research grade
3-(4-Bromophenyl)thiophene


Formula: C₁₀H₇BrS
MW: 239.132 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06740295
CAS Number: 20608-85-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty