Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzoxy)-5-chlorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34912-1G 34912-25G 34912-500MG 34912-5G
76915-258EA 155.96 USD InStock
76915-258 76915-260 76915-262 76915-264
3-(Benzoxy)-5-chlorobenzoic acid ≥97% research grade
3-(Benzoxy)-5-chlorobenzoic acid


Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24038800
CAS Number: 53985-44-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty