Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluoro-6-iodobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34955-1G 34955-25G 34955-500MG 34955-5G
76915-272EA 194.15 USD InStock
76915-272 76915-274 76915-276 76915-278
3-Chloro-2-fluoro-6-iodobenzoic acid ≥97% research grade
3-Chloro-2-fluoro-6-iodobenzoic acid


Formula: C₇H₃ClFIO₂
MW: 300.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31805781
CAS Number: 1252989-58-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty