Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorophenyl)-5-methoxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

35126-1G 35126-25G 35126-500MG 35126-5G
76915-354EA 379.81 USD InStock
76915-354 76915-356 76915-358 76915-360
3-(4-Fluorophenyl)-5-methoxypyridine ≥97%
3-(4-Fluorophenyl)-5-methoxypyridine


Formula: C₁₂H₁₀FNO
Storage Temperature: Refrigerator
MDL Number: MFCD14699909
CAS Number: 1214374-19-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty