Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylthio)benzaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

36325-1G 36325-25G 36325-5G
76915-578EA 91.24 USD InStock
76915-578 76915-580 76915-582
3-(Methylthio)benzaldehyde ≥95%
3-(Methylthio)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈OS
MW: 152.213 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09864229
CAS Number: 73771-35-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty