Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',5'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37099-1G 37099-25G 37099-5G
76915-714EA 106.09 USD InStock
76915-714 76915-716 76915-718
3',5'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade
3',5'-Dimethyl-[1,1'-biphenyl]-4-carboxylic acid


Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201436
CAS Number: 505082-92-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty