Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoro-5-formylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37633-1G 37633-500MG 37633-5G
76915-794EA 498.63 USD InStock
76915-794 76915-796 76915-798
3,4-Difluoro-5-formylbenzoic acid ≥97% research grade
3,4-Difluoro-5-formylbenzoic acid


Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707039
CAS Number: 2386498-20-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty