Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-2-fluorophenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

37724-1G 37724-25G 37724-500MG 37724-5G
76915-808EA 263.11 USD InStock
76915-808 76915-810 76915-812 76915-814
4-(Benzyloxy)-2-fluorophenol ≥97% research grade
4-(Benzyloxy)-2-fluorophenol


Formula: C₁₃H₁₁FO₂
MW: 218.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23110935
CAS Number: 84352-10-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty