Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclobutylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

38225-1G 38225-500MG 38225-5G
76915-938EA 558.04 USD InStock
76915-938 76915-940 76915-942
3-Cyclobutylbenzoic acid ≥97% research grade
3-Cyclobutylbenzoic acid


Formula: C₁₁H₁₂O₂
MW: 176.212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20651608
CAS Number: 19936-19-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty