Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-5-chloro-2-fluoropyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

38731-1G 38731-25G 38731-500MG 38731-5G
76915-992EA 173.99 USD InStock
76915-992 76915-994 76915-996 76915-998
3-(Benzyloxy)-5-chloro-2-fluoropyridine ≥97% research grade
3-(Benzyloxy)-5-chloro-2-fluoropyridine


Formula: C₁₂H₉ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD32902663
CAS Number: 1616667-42-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty