Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-fluorophenylboronic acid hydrochloride ≥95% research grade

Supplier: AOB CHEM USA
Warning

39313-1G 39313-25G 39313-5G
76916-024EA 84.88 USD InStock
76916-024 76916-026 76916-028
4-Amino-3-fluorophenylboronic acid hydrochloride ≥95% research grade
4-Amino-3-fluorophenylboronic acid hydrochloride


Formula: C₆H₇BFNO₂·HCl
MW: 191.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11867880
CAS Number: 1256355-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty