Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-methylthiophenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

39937-1G 39937-25G 39937-500MG 39937-5G
76916-104EA 257.8 USD InStock
76916-104 76916-106 76916-108 76916-110
3-Bromo-5-methylthiophenylboronic acid pinacol ester ≥97% research grade
3-Bromo-5-methylthiophenylboronic acid pinacol ester


Formula: C₁₃H₁₈BBrO₂S
Storage Temperature: Ambient
MDL Number: MFCD16618966
CAS Number: 1256359-20-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty