Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-5-methoxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

45432-1G 45432-25G 45432-500MG 45432-5G
76916-326EA 213.25 USD InStock
76916-326 76916-328 76916-330 76916-332
4-Bromo-2-chloro-5-methoxypyridine ≥97%
4-Bromo-2-chloro-5-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClNO
MW: 222.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09972186
CAS Number: 1020253-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty