Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-4-fluorophenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

45536-1G 45536-25G 45536-500MG 45536-5G
76916-358EA 272.66 USD InStock
76916-358 76916-360 76916-362 76916-364
3-Ethoxy-4-fluorophenol ≥97% research grade
3-Ethoxy-4-fluorophenol


Formula: C₈H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD29044441
CAS Number: 1443055-75-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty