Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-butoxy-5-methylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

45927-1G 45927-25G 45927-500MG 45927-5G
76916-390EA 379.81 USD InStock
76916-390 76916-392 76916-394 76916-396
3-Bromo-2-butoxy-5-methylbenzoic acid ≥97% research grade
3-Bromo-2-butoxy-5-methylbenzoic acid


Formula: C₁₂H₁₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD32707146
CAS Number: 2413441-20-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty