Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorobenzyloxy)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

46027-1G 46027-25G 46027-500MG 46027-5G
76916-416EA 144.29 USD InStock
76916-416 76916-418 76916-420 76916-422
4-(4-Fluorobenzyloxy)benzoic acid ≥97%
4-(4-Fluorobenzyloxy)benzoic acid


Formula: C₁₄H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD04510035
CAS Number: 405-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty