Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-2,6-difluorobenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

48002-1G 48002-25G 48002-500MG 48002-5G
76916-566EA 253.56 USD InStock
76916-566 76916-568 76916-570 76916-572
3,5-Dibromo-2,6-difluorobenzoic acid ≥95%
3,5-Dibromo-2,6-difluorobenzoic acid


Formula: C₇H₂Br₂F₂O₂
Storage Temperature: Ambient
CAS Number: 28314-84-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty