Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-iodobenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

51210-1G 51210-25G 51210-500MG 51210-5G
76916-802EA 173.99 USD InStock
76916-802 76916-804 76916-806 76916-808
3-Fluoro-5-iodobenzaldehyde ≥97% research grade
3-Fluoro-5-iodobenzaldehyde


Formula: C₇H₄FIO
MW: 250.009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779030
CAS Number: 914636-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty