Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzylthio)benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

53059-1G 53059-25G 53059-500MG 53059-5G
76916-958EA 215.37 USD InStock
76916-958 76916-960 76916-962 76916-964
4-(Benzylthio)benzaldehyde ≥97% research grade
4-(Benzylthio)benzaldehyde


Formula: C₁₄H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD18887727
CAS Number: 78832-95-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty